Safranine O solution 0,6%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: CI 50240

M-Clarity™ quality level = MQ500

HT90432-1L
HT90432-1LEA 1020 NOK
HT90432-1L
Safranine O solution 0,6%, Sigma-Aldrich®
Safranine O

Intended for use in Sigma Gram Stain Procedure, HT90.


Sertifiseringer: Certified safranin 0.6% in 20% ethyl alcohol, SD3A.

Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Flash punkt: 37.8 °C (closed cup)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Red
Appearance (Form) Solution
Appearance (Turbidity)
Essentially free of particles/precipitate Clear
Performance Conforms
Gram positive organisms are purple-black in both bacteriology smears and tissue sections; Gram negative organisms are red in both bacteriology smears and tissue sections.
Expiration Date Period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR