Enclosed General Purpose Trolley
Leverandør: HUPFER METALLWERKE
0112054
HUPF0112054EA 0 NOK
HUPF0112054
Enclosed General Purpose Trolley
Traller
Trolley in robust, self-supporting and hygienic design, made of high-quality stainless steel. Ø 25 mm round tube frame with two welded, 1000 ×600 mm deep-drawn shelves with 10 mm high profile edge, lined with sound insulation on the underside, hygienically folded by 35 mm and rebated inwards, designed for easy cleaning. Push bars integrated on both sides into tube frame ensure good manoeuvrability. Trolley with stainless steel panels on three sides and double-wing doors with grip opening and magnetic catch. Four polypropylene disc bumpers serve as bumpers and protect trolley on all sides asMore Product Information