Clearing Trolley, 3 Shelves
Leverandør: HUPFER METALLWERKE
0127772
HUPF0127772EA 16875 NOK
HUPF0127772
Clearing Trolley, 3 Shelves
Traller
Trolley in robust, self-supporting and hygienic design, made of high-quality stainless steel.
  • Round tube frame with welded
  • Deep-drawn shelves with profile edge
  • Lined with sound insulation on the underside
  • Hygienically folded and rebated inwards
  • Designed for easy cleaning
Uppermost shelf with welded, 3-sided stainless steel apron. Push bars integrated on both sides into tube frame ensure good manoeuvrability. 4 polyethylene disc bumpers serve as bumpers and protect trolley on all sides as well as building-side walls from being damaged. Trolley runs on 4More Product Information