Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Leverandør: HyClone products (Cytiva)
SH30268.01 SH30031.02 SH30015.02 SH30015.03 SH30016.02 SH30030.02 SH30031.01 SH30031.03 SH30031.FS SH30268.02 SH30268.LS SH30588.01 SH30588.02
HYCLSH30268.01EA 355 NOK
HYCLSH30268.01 HYCLSH30031.02 HYCLSH30015.02 HYCLSH30015.03 HYCLSH30016.02 HYCLSH30030.02 HYCLSH30031.01 HYCLSH30031.03 HYCLSH30031.FS HYCLSH30268.02 HYCLSH30268.LS HYCLSH30588.01 HYCLSH30588.02
Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Buffere Buffere, cellekultur
Balanced salt solutions for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Powder formats rehydrate to specified quantity
  • Solutions: 0,1 µm sterile filtered
Buffer cell culture growth with HyClone Hank's Balanced Salt Solutions (HBSS), available in lot sizes that facilitate scale-up and manufacturing.