Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid, Cytiva
Leverandør: HyClone products (Cytiva)
SH30025.01
HYCLSH30025.01EA 200 NOK
HYCLSH30025.01
Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid, Cytiva
Media, Cellekultur
Ham's F-10 was designed for use with Chinese Hamster Ovary (CHO) cells, and has been shown to support the growth of human diploid cells, white blood cells, and primary explants from rat, rabbit, or chicken tissues. This classical media can be used for serum-free growth of some cell lines including CHO cells. Serum supplementation may or may not be necessary depending on the type of cell being cultured. Ham's F-10 contains a wide variety of components, including zinc, hypoxanthine, and thymidine and is buffered with sodium bicarbonate.
  • Contains 6,1 mM glucose
  • All liquid media products are hydrated using WFI quality water
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes
A Certificate of Analysis is included in each package with preparation instructions, intended use, and handling and performance criteria.
Bestillingsinformasjon: Liquid media are supplied in unbreakable, square PETG bottles.