Natriumhydrogenkarbonat 75 g/l vandig løsning, HyClone™, sterilfiltrert

Leverandør: HyClone products (Cytiva)

SH30033.01
HYCLSH30033.01EA 280 NOK
HYCLSH30033.01
Natriumhydrogenkarbonat 75 g/l vandig løsning, HyClone™, sterilfiltrert
Natriumhydrogenkarbonat

Buffer cell culture with a fully QC-tested solution.


  • 0.1 µm sterile filtered
  • pH 7.9 to 9.4
  • Endotoxin Concentration: ≤1.0 EU/mL


This solution has full traceability and documented origin of all formula ingredients. It is validated and consistent for production processes.


Sertifiseringer: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

Formel: NaHCO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003528
CAS-nummer: 144-55-8
EINECS: 205-633-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Endotoxin concentration ≤1.0 EU/mL
Sterilization 0.1 µm sterile filtered
pH 7.9 - 9.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR