Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Leverandør: HyClone products (Cytiva)
SH30212.03
HYCLSH30212.03EA 2500 NOK
HYCLSH30212.03
Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Serum
HyClone™ Alpha Calf Fraction provides hybridoma users with a cost-effective medium supplement that promotes growth and antibody production.