Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Leverandør: HyClone products (Cytiva)
SH30256.01 SH30258.01 SH30256.02 SH30256.FS SH30256.LS SH30258.02
SH30256.01EA 350 NOK
SH30256.01 HYCLSH30258.01 HYCLSH30256.02 HYCLSH30256.FS HYCLSH30256.LS HYCLSH30258.02
Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Buffere Buffere, cellekultur
Balanced salt solution used for multiple cell culture applications.
  • Full traceability and documented origin of all formula ingredients
  • Validated and consistent production processes
0,1 µm sterile filtered
Endotoxin Concentration: ≤1.0 EU/mL
Sertifiseringer: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities