CHO cell culture platform media, HyClone™ SFM4CHO Utility
Leverandør: HyClone products (Cytiva)
SH30517.01
HYCLSH30517.01EA 1950 NOK
HYCLSH30517.01
CHO cell culture platform media, HyClone™ SFM4CHO Utility
Media, Cellekultur
HyClone™ SFM4CHO-Utility media is ideally suited for the cost-effective manufacturing of recombinant proteins for academic and industrial research, genomics and proteomics, drug target screening and validation, and manufacturing of preclinical lots. It is formulated using a proprietary lipid complexing process for enhanced stability and broad-spectrum growth promotion.
  • Supports the DHFR selection/amplification system
  • Defined protein-free formulation
  • Metabolically designed for high cell yield and recombinant protein production
  • Supports multiple CHO clones
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
SFM4CHO-Utility does not contain phenol red. It has been designed to support the DHFR selection/amplification system. This protein-free media has been successfully tested in a variety of culture systems including T-flasks, spinner and shaker flasks, roller bottles, and bioreactors.