L-alanyl L-glutamine, SG-200, HyClone™
Leverandør: HyClone products (Cytiva)
SH30590.01
HYCLSH30590.01EA 140 NOK
HYCLSH30590.01
L-alanyl L-glutamine, SG-200, HyClone™
Cellekultursupplementer og tilsetninger
SG-200 solution is a stable dipeptide of L-alanyl L-glutamine.