Screw vials, Thermo Scientific™ Chromacol™
Leverandør: Thermo Fisher Scientific
1.1-STVGN
HYPE1.1-STVGNEA 6975 NOK
HYPE1.1-STVGN
Screw vials, Thermo Scientific™ Chromacol™
Vialer
Thermo Scientific™ Chromacol™ sample storage screw thread vials are manufactured from superior quality 33 expansion borosilicate glass, clear (Type 1, Class A).
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.