SPE plates, 96-well, HyperSep™, SAX
Leverandør: Thermo Fisher Scientific
60300-567
HYPE60300-567EA 6675 NOK
HYPE60300-567
SPE plates, 96-well, HyperSep™, SAX
Mikroplater Solid fase ekstaksjonsplater
Thermo Scientific™ HyperSep SAX Strong Anion Exchanger SPE 96-Well Plates and Individual Wells feature a strong anion exchange sorbent for extraction of weak acids. The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.