HPLC Columns, μPAC™ Neo
Leverandør: Thermo Fisher Scientific
COL-NANO110NEO
HYPECOL-NANO110NEOEA 0 NOK
HYPECOL-NANO110NEO
HPLC Columns, μPAC™ Neo
Kromatografikolonner
μPAC Neo column line offers higher resolution ensuring that you can extract the maximum amount of information from highly complex samples using low-flow chromatography.
  • Operating at lower back pressures extends column lifetime
  • Designed with a perfect flow path order, eliminating heterogeneous flow paths
The μPAC Neo column range is the ideal choice for a variety of proteomics experiments, including fast label-free quantitation, comprehensive identification, and single cell analysis.
These columns operate at moderate pressures, allowing for longerMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Kappet ved endene Ja
Maks. trykk 450 bar
pH område 1,5 - 7
Temperaturgrense 60 °C