μPAC™ ESI-MS Accessories
Leverandør: Thermo Fisher Scientific
EMI-EASYSPRAYB
HYPEEMI-EASYSPRAYBEA 15300 NOK
HYPEEMI-EASYSPRAYB
μPAC™ ESI-MS Accessories
Kromatografiutstyr Kromatografireservedeler
ESI-MS accessories are used to facilitate reproducible, low dispersion connections between μPAC HPLC columns and low-flow mass spectrometer ion sources.
The μPAC EASY-spray emitter is equipped with a stainless steel 50 μm through-bore union. Achieve high resolution separations in low-flow applications when using this emitter to connect a μPAC HPLC column and a transfer line with a EASY-Spray™ source.
The μPAC EASY-spray emitter is designed for easy connection to an EASY-Spray MS source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.