Screw caps with valves, Mininert
Leverandør: Thermo Fisher Scientific
TS-10130 TS-10135
HYPETS-10130EA 3550 NOK
HYPETS-10130 HYPETS-10135
Screw caps with valves, Mininert
Lokk Lokk til vials
Mininert valves are screw caps that have integrated resealable valves to allow repeated and unlimited syringe-needle access to samples eliminates the septum-boring that occurs with repeated puncture of traditional septa. Made of chemical-resistant PTFE coating, they provide an inert, leak-tight, high-pressure seal.