Small reaction vials, Reacti-Vial™
Leverandør: Thermo Fisher Scientific
TS-13225 TS-13100 TS-13220 TS-13221 TS-13097 TS-13223 TS-13222 TS-13099
HYPETS-13225EA 5575 NOK
HYPETS-13225 HYPETS-13100 HYPETS-13220 HYPETS-13221 HYPETS-13097 HYPETS-13223 HYPETS-13222 HYPETS-13099
Small reaction vials, Reacti-Vial™
Vialer
These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.
  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds
These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solventMore Product Information