Siliconising Fluid, water soluble
Leverandør: Thermo Fisher Scientific
TS-42799
HYPETS-42799EA 3025 NOK
HYPETS-42799
Siliconising Fluid, water soluble
Siliconising Fluid
This multifunctional siliconising fluid is specially designed to chemically bind microscopically thin, water-repellent films to glass, quartz, silica and ceramics. The coated surfaces are neutral, hydrophobic and non-oily.
  • Components: 4-Hydroxy-4-methylpentan-2-on, 2-Methyl-2-Propanol, Silan, Trimethoxyoctadecyl
  • Concentrations: 25 – 45, 25 – 45, 10 – 20 %
This easy-to-use silane monomer solution is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol. The primary silane component is an octadecyltrialkoxysilane, which is hydrolyzed to a silanol when mixed with water. It provides greater resistance to base hydrolysis than other surface treatments.