Silika gel, Silitech

Leverandør: MP Biomedicals

Synonymer: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

0202071
ICNA0202071EA 6625 NOK
ICNA0202071
Silika gel, Silitech
Silika gel

Mean Pore Size: 60Å
Particle Size: 32-63 µm

Formel: SiO₂
Tetthet: ∼2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 7631-86-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR