5-Bromo-2′-deoxycytidine hydrate, white powder

Leverandør: MP Biomedicals

0210016680 0210016683 0210016690 0210016691
ICNA0210016680EA 890 NOK
ICNA0210016680 ICNA0210016683 ICNA0210016690 ICNA0210016691
5-Bromo-2′-deoxycytidine hydrate, white powder
5-Bromo-2′-deoxycytidine hydrate

5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP.


Soluble in water, NH₄OH (50 mg /ml).

Formel: C₉H₁₂BrN₃O₄·×H₂O
Lagringstemperatur: Fryseskap
CAS-nummer: 1022-79-3

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
FTIR Conforms to Standard
UV/VIS: λmax (water) 286 - 289 nm
Moisture Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR