Adonitol, white powder

Leverandør: MP Biomedicals

0210025205 0210025225 0210025280
ICNA0210025205EA 675 NOK
ICNA0210025205 ICNA0210025225 ICNA0210025280
Adonitol, white powder
Adonitol

Adonitol (ribitol), a pentose alcohol, is metabolised to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.


Adonitol is a crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution.


Soluble in water (100 mg/ml - clear, colorless solution)


NB: Research Use Only (RUO).

Formel: HOH₂C(CH(OH))₃CH₂OH
MDL nummer: MFCD00064291
CAS-nummer: 488-81-3

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Melting Point 98 - 106 °C
FTIR Conforms to Standard
Moisture Content < 0.50%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR