Alizarin red S (sodium salt), anhydrous, orange powder C.I. 58005

Leverandør: MP Biomedicals
Warning

Synonymer: C.I. 58005

0210037590 0210037525 0210037580
ICNA0210037590EA 14600 NOK
ICNA0210037590 ICNA0210037525 ICNA0210037580
Alizarin red S (sodium salt), anhydrous, orange powder C.I. 58005
Alizarin red S (sodium salt)

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Soluble in water (1 mg/ml).

Formel: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR