Penicillin G kaliumsalt

Leverandør: MP Biomedicals
Warning

0210054325 0210054380 0210054310
ICNA0210054325EA 840 NOK
ICNA0210054325 ICNA0210054380 ICNA0210054310
Penicillin G kaliumsalt
Penicillin G kaliumsalt

Penicillin G is a narrow spectrum antibiotic.


Penicillin G is active against gram-positive and gram-negative aerobic and anaerobic bacteria. Penicillin G inhibits cell wall synthesis by binding to penicillin binding proteins (PBPs) which inhibits peptidoglycan chain cross-lining. Penicillin G is stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases.


It is the drug of choice for Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus. Penicillin G potassium salt is used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.


Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking..


Soluble in water (100 mg/ml) clear,colourless to light yellow.

Formel: C₁₆H₁₇KN₂O₄S
Smeltepunkt: 214…217 °C
Tetthet: 1,359 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036193
CAS-nummer: 113-98-4
EINECS: 204-038-0
Merck Indeks: 12,07225

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR