Choline hydrogen tartrate ≥98%, white powder

Leverandør: MP Biomedicals
Warning

Synonymer: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

0210138483 0210138490
ICNA0210138483EA 1050 NOK
ICNA0210138483 ICNA0210138490
Choline hydrogen tartrate ≥98%, white powder
Choline hydrogen tartrate

Choline bitartrate is a acyl group acceptor. Bitarate is added to the choline to speed up the absorption rate within the human body. Choline Bitartrate is an essential nutrient, generally grouped as a B vitamin, that helps to improve cognitive function and aids fat metabolism.


  • Acyl group acceptor
  • Presentation: White Powder


Choline bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.


Soluble in alcohol; slightly soluble in water (100 mg/ml in H₂O).


NB: Research Use Only (RUO).

Formel: C₉H₁₉NO₇
Smeltepunkt: 146…155 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036332
CAS-nummer: 87-67-2

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (2% aq soln) 3 - 4
Loss on Drying ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR