Folic acid ≥95%, orange powder

Leverandør: MP Biomedicals

Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

0210172505 0210172525 0210172580
ICNA0210172505EA 525 NOK
ICNA0210172505 ICNA0210172525 ICNA0210172580
Folic acid ≥95%, orange powder
Folic acid

Folic acid, also known as folate, is a B vitamin that can be found in a variety of fruits and vegetables. It can also be chemically synthesized. Folate, a watersoluble vitamin, helps the body form red blood cells and aids in the formation of genetic material within every body cell. This product exhibits metal binding properties. Hematopoietic vitamin present, free or combined with one or more additional molecules of L-(+)-glutamic acid, in liver, kidney, mushrooms, spinach, yeast, green leaves, and grasses.
Folic acid (Vitamin B9 and folate) is essential to numerous bodily functions. The human body needs folate to synthesize DNA, repair DNA, and methylate DNA as well as to act as a cofactor in certain biological reactions. It is especially important in aiding rapid cell division and growth, such as in infancy and pregnancy.
Folic acid (FA) and dihydrofolic acid (FAH2) are substrates of dihydrofolate reductase(s) which reduce them to tetrahydrofolate (THF), which in turn supports ‘one carbon’ transfer. Tetrahydrofolates are required for de novo synthesis of purines, thymidylic acid and various amino acids and for post-translational methylation (epigenetics).
Very slightly soluble in cold water (0.0016 mg/mL at 25 °C), soluble to about 1% in boiling water. Slightly soluble in methanol, appreciably less soluble in ethanol and butanol. Insoluble in acetone, chloroform, ether, benzene. Relatively soluble in acetic acid, phenol, pyridine, solutions of alkali hydroxides and carbonates. Soluble in hot dilute HCl and H2SO4.


  • Presentation: Orange Powder
  • Extinction Coefficient (E1%): 9.12, 25.1, 26.9 (0.1 M NaOH)(Lit.)
  • A suspension of 1 g folic acid in 10 mL water has a pH of 4.0-4.8. Aqueous solutions prepared with sodium bicarbonate have a pH between 6.5 and 6.8.
  • pKa: 8.26 (Lit.)
  • Storage temperature: Store at Room Temperature (15-30 °C). Protect from light.

Formel: C₁₉H₁₉N₇O₆
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00079305
CAS-nummer: 59-30-3

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥95.0%
FTIR Conforms to Standard
UV/Vis: l max 256 ± 3 nm (0.1 N NaOH)
Loss on Drying ≤8.5%
Residue on Ignition ≤0.3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR