(S)-(-)-Malein syre ≥99%, pulver

Leverandør: MP Biomedicals
Warning

0210223790 0210223791 0210223725 0210223780
ICNA0210223790EA 2800 NOK
ICNA0210223790 ICNA0210223791 ICNA0210223725 ICNA0210223780
(S)-(-)-Malein syre ≥99%, pulver
(S)-(-)-Malein syre

L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.


L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.


Soluble in 100 mg/ml - clear to very slightly hazy, colourless.

Formel: C₄H₆O₅
Kokepunkt: 322 °C (1013 hPa)
Smeltepunkt: 101…104 °C
Tetthet: 1,595 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064213
CAS-nummer: 97-67-6
EINECS: 202-601-5
Merck Indeks: 13,05730

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR