L-Metionin

Leverandør: MP Biomedicals

02102291.1 02102291.5 0210229101 02102291.3
ICNA02102291.1EA 970 NOK
ICNA02102291.1 ICNA02102291.5 ICNA0210229101 ICNA02102291.3
L-Metionin
L-Metionin
Formel: C₅H₁₁NO₂S
Kokepunkt: 278 °C (1013 hPa)
Smeltepunkt: 136 °C
Tetthet: 1,373 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063097
CAS-nummer: 63-68-3
EINECS: 200-562-9
Merck Indeks: 13,06004

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR