Theophyllin, vannfri, white powder

Leverandør: MP Biomedicals
Danger

0210302480 0210302483 0210302490 0210302491
ICNA0210302480EA 675 NOK
ICNA0210302480 ICNA0210302483 ICNA0210302490 ICNA0210302491
Theophyllin, vannfri, white powder
Theophyllin

Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.


Soluble in 0,1 M HCl; 0,1 M NaOH; slightly soluble in water; moderately soluble in ethanol. Soluble in 1 M NH4OH (50 mg/ml - clear colorless solution) and 50% ammonium hydorxide (50 mg/ml - clear, colorless solution).


NB: Research Use Only (RUO).

Formel: C₇H₈N₄O₂
MDL nummer: MFCD00079619
CAS-nummer: 58-55-9
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
FTIR Conforms to Standard

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR