Tris-(hydroksymetyl)aminometan hydroklorid ≥99%, Reagent Grade

Leverandør: MP Biomedicals

02103130.2 02103130.1 02103130.5 0210313001 0210313005
ICNA02103130.2EA 1225 NOK
ICNA02103130.2 ICNA02103130.1 ICNA02103130.5 ICNA0210313001 ICNA0210313005
Tris-(hydroksymetyl)aminometan hydroklorid ≥99%, Reagent Grade
Tris-(hydroksymetyl)aminometan hydroklorid

Tris and Tris Hydrochloride have been useful as buffers in a wide variety of biological systems. Uses include pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.


Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes). It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid .


The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0,03 unit per °C decrease in temperature, and decreases 0,03-0,05 unit per ten-fold dilution.

Formel: NH₂C(CH₂OH)₃·HCl
Smeltepunkt: 150…152 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012590
CAS-nummer: 1185-53-1
EINECS: 214-684-5
Merck Indeks: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR