Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Leverandør: MP Biomedicals
0210330401
ICNA0210330401EA 865 NOK
ICNA0210330401
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Mikrobiologiske medier Base Materialer for Mikrobiologi
Yeast Hydrolysate Enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components, carbohydrates, and B complex vitamins natural to yeast.
  • Soluble in water
  • Standard plate count: <50,000/g
  • Protein (N×6,25): 50,0 to 65,0%