Kloramin T trihydrat, white crystalline powder

Leverandør: MP Biomedicals
Danger

Synonymer: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

15011591 0215011583 0215011591
ICNA15011591EA 830 NOK
ICNA15011591 ICNA0215011583 ICNA0215011591
Kloramin T trihydrat, white crystalline powder
Kloramin T trihydrat

Soluble in water (100 mg/ml); practically insoluble in benzene, chloroform or ether.


  • Presentation : White Crystalline Powder
  • pH : 6 - 9(1% aq soln)
  • Detection Method : FTIR
  • Solubility :


Storage: protect from light, under nitrogen.

Formel: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Smeltepunkt: 167...170 °C (dec., lit.)
Tetthet: 540...680 kg/m³ (bulk density)
Flash punkt: 192 °C (closed cup)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00149066
CAS-nummer: 7080-50-4
EINECS: 204-854-7
FN: 3263
ADR: 8 III
Merck Indeks: 13,02085

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 8 - 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR