Poly-L-lysine hydrobromide, MP Biomedicals

Leverandør: MP Biomedicals

0215017710 0215017725 0215017750 0215017780
ICNA0215017710EA 575 NOK
ICNA0215017710 ICNA0215017725 ICNA0215017750 ICNA0215017780
Poly-L-lysine hydrobromide, MP Biomedicals
Proteiner

Poly-L-Lysine is typically used as a coating substrate for culture dishes, slides, etc.


It can be used in conjugation to methotrexate for increased drug transport. For Microencapsulation of islets. It is also use in simple reproducible procedure for chromosomal preparations from a variety of tissues. For immobilation of living cells in biocompatible semipermeable microcapsules. In the preparation of polycationic beads. Conjugation to albumin and horseradish peroxidase to enhance cellular uptake.


Poly-lysine with mol. wt. >70,000 is useful in promoting cell adhesion to solid substrates.


All molecular weights are calculated from the viscosity of the parent compound Poly-epsilon-Z-L-Lysine.


Soluble in water (50 mg/ml). Dissolve 10 mg poly-L-lysine in 1 ml sterile, deionised water. This creates a 1% solution. Effective coating solution concentration for culture dishes will be 0,3 to 0,5% and for histology slides at 0,1%.


NB: Research Use Only (RUO).

Bestill nå

SPESIFIKASJONER

Protein/peptide name Poly-L-lysine hydrobromide
Environmentally Preferable
Arter
Kilde
CAS-nr 25988-63-0
MDL No. MFCD00237230
Skjema Cream-coloured Powder
Molekylvekt ≥70,000 kDa
Molekylformel (C₆H₁₃N₂OBr)n
Lagringsforhold Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR