Azure B (azure I), brown powder C.I. 52010

Leverandør: MP Biomedicals

Synonymer: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

0215041705 0215041701
ICNA0215041705EA 1250 NOK
ICNA0215041705 ICNA0215041701
Azure B (azure I), brown powder C.I. 52010
Azure B (azure I)

Azure B is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Soluble in water (50 mg/ml) or ethanol (8-10 mg/ml); insoluble in diethyl ether.

Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Smeltepunkt: 205…210 °C (dec., lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011935
CAS-nummer: 531-55-5
EINECS: 208-511-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR