Brilliant cresyl blue, dark green powder C.I. 51010

Leverandør: MP Biomedicals
Warning

0215051525 0215051505 0215051580
ICNA0215051525EA 2350 NOK
ICNA0215051525 ICNA0215051505 ICNA0215051580
Brilliant cresyl blue, dark green powder C.I. 51010
Brilliant cresyl blue

Brilliant Cresyl Blue is a supravital stain used in hematology for demonstrating and counting reticulocytes and platelets. It is also used as a vital stain for demonstrating the mucous secretions of the marine coelomates.


Satisfactory for staining reticulocytes in fresh blood using both aqueous and alcoholic stain preparations.


Soluble in water, 1 N Hydrochloric acid, or 50% ethanol.

Formel: C₃₄H₄₂Cl₄N₈O₂Zn
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00148901
CAS-nummer: 81029-05-2
EINECS: 225-203-3

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
UV/Vis: l max (ethanol) For Information Only

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR