Eucalyptol ~99%, clear, colourless liquid

Leverandør: MP Biomedicals
Warning

0215068990 0215068950
ICNA0215068990EA 0 NOK
ICNA0215068990 ICNA0215068950
Eucalyptol ~99%, clear, colourless liquid
Eucalyptol

Eucalyptol (cineole) is an ingredient in many brands of mouthwash and cough suppressant. It is used as an insecticide and insect repellent.It is used in flavorings, fragrances, and cosmetics.


Soluble in alcohol, most fixed oils, organic solvents, glycerin, propylene glycol, chloroform and ether; insoluble in water.

Formel: C₁₀H₁₈O
Kokepunkt: 174…177 °C
Tetthet: 0,921…0.924 g/cm³ (20 °C)
Flash punkt: 50 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167977
CAS-nummer: 470-82-6
EINECS: 207-431-5
FN: 1993
ADR: 3,III
Merck Indeks: 12,03940

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR