Fura 2-AM, solid

Leverandør: MP Biomedicals

Synonymer: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

02152286.5 0215228601
ICNA02152286.5EA 3900 NOK
ICNA02152286.5 ICNA0215228601
Fura 2-AM, solid
Fura 2-AM

The acetoxymethyl ester of the chelator is soluble in dimethyl sulfoxide (10 mg/ml) and in nonpolar solvents such as chloroform, acetone and benzene. It is insoluble in water or methanol.


  • Ion Specificity: Ca2+ > Mg2+, protons
  • Dissociation constants: Ca2+ complex - 135 - 224 nM; Mg2+ complex - 6 - 10 mM

Formel: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
MDL nummer: MFCD00036976
CAS-nummer: 108964-32-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR