Kalium heksacyanoferrat (II) trihydrat 98.5-102.0%, yellow crystalline powder ACS

Leverandør: MP Biomedicals

0215256080 0215256090
ICNA0215256080EA 535 NOK
ICNA0215256080 ICNA0215256090
Kalium heksacyanoferrat (II) trihydrat 98.5-102.0%, yellow crystalline powder ACS
Kalium heksacyanoferrat (II) trihydrat

Potassium ferrocyanide trihydrate is a salt of the Fe(CN)₆4- ferrocyanide ion. Ferrocyanide is a reducing iron species, being oxidised from ferrous Fe(II) to ferric Fe(III).


It is used to determine the concentration of potassium permanganate, a compound often used in titrations based on redox reactions. Used in a mixture with potassium ferricyanide and phosphate buffered solution (PBS) to provide a buffer for X-Gal, which is used to cleave Beta-galactosidase, giving a bright blue visualisation where an antibody (or other molecule), conjugated to Beta-gal, has bonded to its target.


It is membrane impermeable and can be used to assess redox activity of membrane-bound structures and mechanisms of electron transport across membranes. This approach was employed to demonstrate that Potassium ferrocyanide performs reducing activation of membrane-bound dopamine β-monooxygenase from the exterior face of adrenal vesicles, providing elucidation of native reduction-activation mechanisms for dopamine β-monooxygenase regulation.

Formel: K₄Fe(CN)₆·3H₂O
Smeltepunkt: 70 °C (lit.)
Tetthet: 7,87 g/cm³ (25 °C) (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167023
CAS-nummer: 14459-95-1

Bestill nå

Spesifikasjon Testresultater

Assay 98,5 - 102,0%
Insoluble Matter ≤0,005%
Chloride ≤0,01%
Sulphate Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR