Indigo carmine, dark powder

Leverandør: MP Biomedicals
Warning

Synonymer: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

0215263805 0215263880
ICNA0215263805EA 415 NOK
ICNA0215263805 ICNA0215263880
Indigo carmine, dark powder
Indigo carmine

Indigo Carmine is a blue, anionic indigoid dye.
Indigo Carmine is useful in the staining of Negri bodies. It can be used as a contrast stain for plasma when red nuclear stains are used.


  • Presentation: Dark Powder
  • Soluble in water (3 mg/mL) and ethanol (3 mg/mL).
  • Storage temperature: Store at room Temperature, protect from light.

Formel: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
EINECS: 212-728-8
Merck Indeks: 13,04969

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
UV/Vis: l max (water) 605-615nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR