Toluidinblått O, powder C.I. 52040

Leverandør: MP Biomedicals

Synonymer: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

0215264925
ICNA0215264925EA 1625 NOK
ICNA0215264925
Toluidinblått O, powder C.I. 52040
Toluidinblått O

Toluidine blue O is useful as a metachromatic nuclear stain.


Soluble in water (10 mg/ml, blue violet solution), ethanol (4 mg/ml), alcohol (blue solution).

Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
MDL nummer: MFCD00011934
CAS-nummer: 92-31-9
Merck Indeks: 12,09658

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
UV/VIS: λmax (H₂O) 630 ± 4 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR