Alphazurine A, powder C.I. 42080

Leverandør: MP Biomedicals

Synonymer: Fenazo bluexg, Patent Blue A, Carmine Blue AF, Alphazurine A, Erio Glaucine X, Acid Blue 7

0215473025 0215473080
ICNA0215473025EA 845 NOK
ICNA0215473025 ICNA0215473080
Alphazurine A, powder C.I. 42080
Alphazurine A

Soluble in water (1 mg/mL).

Formel: C₃₇H₃₅N₂NaO₆S₂
MW: 690.805 g/mol
MDL nummer: MFCD00044440
CAS-nummer: 3486-30-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR