Cefazolin Sodium Salt

Leverandør: MP Biomedicals
Danger

0215494680 0215494690 0215494691
ICNA0215494680EA 950 NOK
ICNA0215494680 ICNA0215494690 ICNA0215494691
Cefazolin Sodium Salt
Cefazolin sodium salt

Storage temperature.: +4 °C

Formel: C₁₄H₁₄N₈NaO₄S₃
MDL nummer: MFCD00056883
CAS-nummer: 27164-46-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR