Cefotaxime sodium salt ≥95%, off-white powder

Leverandør: MP Biomedicals
Danger

0215494780 0215494790 0215494791
ICNA0215494780EA 1025 NOK
ICNA0215494780 ICNA0215494790 ICNA0215494791
Cefotaxime sodium salt ≥95%, off-white powder
Cefotaxime sodium salt

Cefotaxime is a cephalosporin, which acts by interference of synthesis of peptidoglycan of the bacterial cell wall. Cefotaxime, a cephalosporin antibiotic, is active against both gram-negative and gram-positive bacteria. In addition, cefotaxime blocks the division of cyanobacteria and lower plant organelles/plastids such as the photosynthetic organelles of Glaucophytes and chloroplasts of bryophytes.
Cefotaxime is used for infections of the respiratory tract, skin, bones, joints, urogenital system, meninges, and bloodstream. It generally has good coverage against most Gram-negative bacteria, with the notable exception of Pseudomonas. It is also effective against most Gram-positive cocci except for Enterococcus. It is active against penicillin-resistant strains of Streptococcus pneumoniae. It has modest activity against the anaerobic Bacteroides fragilis. In meningitis, cefotaxime crosses the blood–brain barrier better than cefuroxime.
Cefotaxim inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs) which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes.


  • Presentation: Off-White Powder
  • Soluble in water (0.05 gm/ 1 mL-clear, light yellow solution).
  • Storage temperature: Store at +4 °C, protect from light


Storage temperature.: +4 °C

Formel: C₁₆H₁₇N₅O₇S₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00079073
CAS-nummer: 64485-93-4

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥95%
FTIR Conforms to Reference

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR