Ionomycin calcium salt

Leverandør: MP Biomedicals

0215507001 0215507005
ICNA0215507001EA 2775 NOK
ICNA0215507001 ICNA0215507005
Ionomycin calcium salt
Ionomycin calcium salt

Ionomycin is an ionophore produced by the bacterium Streptomyces conglobatus.A polyether ionic antibiotic that exhibits an affinity for Ca2+. It is used in research to raise the intracellular level of calcium (Ca2+) and as a research tool to understand Ca2+ transport across biological membranes. These cytokines are important in the inflammatory response.


Ionomycin is also used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2, and IL-4 usually in conjunction with PMA.


In Ionomycin Ca2+ ionophore that is more effective than A23187 as a mobile ion carrier for Ca2+; non-fluorescent; used to study Ca2+ transport across biological membranes; induces apoptotic degeneration of embryonic cortical neurons.


Soluble in acetone, benzene, chloroform (50 mg/ml - clear colorless), DMSO (50 mg/ml - clear colourless), CHCl3 (50 mg/ml - clear colourless) and hexane all clear and colourless.

Formel: C₄₁H₇₀CaO₉
MDL nummer: MFCD00151183
CAS-nummer: 56092-82-1

Bestill nå

Spesifikasjon Testresultater

Form White powder
Storage Store at +4 °C. Protect from light
Source Streptomyces conglobatus


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR