Bly tetraacetat ≥95%, powder

Leverandør: MP Biomedicals
Danger

Synonymer: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

0215518225 0215518290 0215518280
ICNA0215518225EA 850 NOK
ICNA0215518225 ICNA0215518290 ICNA0215518280
Bly tetraacetat ≥95%, powder
Bly tetraacetat

Lead tetraacetate smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.

Formel: (H₃CCOO)₄Pb
MW: 443.38 g/mol
Smeltepunkt: 175…180 °C
Tetthet: ∼2.22 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008693
CAS-nummer: 546-67-8
EINECS: 208-908-0
FN: 1616
ADR: 6.1 III
Merck Indeks: 13,05442

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR