Neohesperidin, off-white powder

Leverandør: MP Biomedicals

Synonymer: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (S)-4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside]

0215581380
ICNA0215581380EA 1125 NOK
ICNA0215581380
Neohesperidin, off-white powder
Neohesperidin

A flavanone glycoside with neuroprotective and antioxidant properties. Does not inhibit oral carcinogenesis in a rat model unlike other citrus flavanones, but does exert a protective effect against gastritis and gastric lesions.


Neohesperidin has been used in a study to assess the composition of shiikuwasha peels of different cultivation lines. It has also been used in a study to investigate the protective effects of an extract from Citrus bergamia against inflammatory injury.


Soluble in pyridine (20 mg/ml - clear, yellow solution).

Formel: C₂₈H₃₄O₁₅
MW: 610,60 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD16661259
CAS-nummer: 13241-33-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR