3-Phenylpropanal ≥95%, clear, colourless liquid

Leverandør: MP Biomedicals
Warning

Synonymer: 3-Phenylpropionaldehyde, Hydrocinnamaldehyde

0215619680 0215619625
ICNA0215619680EA 5875 NOK
ICNA0215619680 ICNA0215619625
3-Phenylpropanal ≥95%, clear, colourless liquid
3-Phenylpropanal

Insoluble in water

Formel: C₉H₁₀O
MW: 134.18 g/mol
Kokepunkt: 224
Smeltepunkt: –42 °C
Tetthet: 1.01 g/cm³ (20 °C)
MDL nummer: MFCD00007021
CAS-nummer: 104-53-0
EINECS: 203-211-8

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥95%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR