Propylenkarbonat, liquid

Leverandør: MP Biomedicals

0215641590 0215641591 0215641594
ICNA0215641590EA 1175 NOK
ICNA0215641590 ICNA0215641591 ICNA0215641594
Propylenkarbonat, liquid
Propylenkarbonat

Soluble in water.


Soluble in water.

Formel: C₄H₆O₃
MDL nummer: MFCD00005385
CAS-nummer: 108-32-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR