Xylen (blanding av isomerer), clear, colourless liquid for histology

Leverandør: MP Biomedicals
Danger

Synonymer: Dimethylbenzene

0215869204
ICNA0215869204EA 1950 NOK
ICNA0215869204
Xylen (blanding av isomerer), clear, colourless liquid for histology
Xylen (blanding av isomerer)

Xylene is used in Giemsa cell staining protocols. Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries.


  • Explosion limit 7% (Lit.)


It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.

Formel: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Kokepunkt: 137…140 °C (lit.)
Smeltepunkt: –47.9…+13.3 °C (lit.)
Tetthet: 0.86 g/mL (25 °C) (lit.)
Flash punkt: 25
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00077264
CAS-nummer: 1330-20-7
FN: 1307
ADR: 3 III
Merck Indeks: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
FTIR Conforms to Standard
Water ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR