Spectinomycin sulphate ≥98%, powder

Leverandør: MP Biomedicals

0215899302
ICNA0215899302EA 970 NOK
ICNA0215899302
Spectinomycin sulphate ≥98%, powder
Spectinomycin sulphate

Spectinomycin sulphate is a broad spectrum antibiotic that inhibits protein synthesis (elongation) by binding to the bacterial 30S ribosomal subunit and interfering with peptidyl tRNA translocation.


Soluble in DMSO, ethanol and water; practically insoluble in pyridine, chloroform,ethyl acetate, cyclohexone, benzene, acetone, DMF, dioxane and acetonitrile.


Store solution aliquoted at –20 °C for up to 3 months.

Formel: C₁₄H₂₄N₂O₇·H₂SO₄
MDL nummer: MFCD00270185
CAS-nummer: 23312-56-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR