Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals

Leverandør: MP Biomedicals

0219012310
ICNA0219012310EA 7600 NOK
ICNA0219012310
Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Enzymer

Yeast lytic enzyme is used to digest the cell wall of active yeast cells. It is prepared from a submerged culture of Arthrobacter luteus which effectively lyses cell walls of viable yeast cell.


  • Used for cell lysis, spheroplasting and glucan hydrolysis
  • Activators: Dithiothreitol, 2-mercaptoethanol and other sulfhydryl compounds (such as cystein)


The primary enzyme in this preparation responsible for lysis of viable yeast cells is β-1,3-glucan laminaripentaohydrolase. It hydrolyzes glucose polymers at the β-1,3-glucan linkages releasing laminaripentaose as the principal product.


Unit definition: one unit will produce a change in absorbance at 800 nm of 0,001 per minute at pH 7,5 and 25 °C, using a suspension of brewers yeast as a substrate in a 3 ml reaction mixture.

Bestill nå

SPESIFIKASJONER

Enzymnavn Yeast lytic enzyme
Environmentally Preferable
Enzym synonymer Zymolyase
Kilde
Arter Arthrobacter luteus
Skjema Off-white powder
Lagringsforhold −20 °C
Enzymaktivitet ≥100,000 u/g
pH 5 - 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR