Ampicillin sodium salt

Leverandør: MP Biomedicals
Danger

0219014805 0219014825 0219014880
ICNA0219014805EA 905 NOK
ICNA0219014805 ICNA0219014825 ICNA0219014880
Ampicillin sodium salt
Ampicillin sodium salt

Ampicillin is a semi-synthetic derivative of penicillin, active as a broad-spectrum antibiotic.


It is inactivated by beta-lactamases and for this reason a beta-lactamase inhibitor should be considered when using ampicillin. Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin; against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines. In E. coli it inhibits cell wall synthesis.


Used to select for ampicillin resistance in mutated and transformed cells.


A β-lactam antibiotic with an amino group side chain attached to the penicillin structure. Penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Bactericidal only to growing Escherichia coli. Mode of resistance: cleavage of β-lactam ring of ampicillin by β-lactamase. Antimicrobial spectrum: gram-negative and gram-positive bacteria.


Soluble in water (50 mg/ml); soluble in 1 M ammonium hydroxide, dilute acids or bases; practically insoluble in alcohol, chloroform, ether and fixed oils.

Formel: C₁₆H₁₈N₃NaO₄S
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00064313
CAS-nummer: 69-52-3

Bestill nå

Spesifikasjon Testresultater

Synonyms D(-)-α-Aminobenzylpenicillin sodium salt
Storage Store at +4 °C
Loss on Drying 2%
pH 8 - 10 (1% aq soln)
pka pKa = 2,5 (-COOH) pKa = 7,3 (-NH2) at 25 °C
Potency 800 μg/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR